• USA Home
  • Search Results

  • sodiumtaurocholatehydrate53768anhydrousbasis34590926411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium taurocholate hydrate

1 Products
Synonym:   Taurocholic acid sodium salt
Image of Sodium taurocholate hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C26H44NNaO7S · xH2O
Molecular Weight:
 537.68 (anhydrous basis)
 Beilstein/REAXYS Number:
 3901620
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More