• USA Home
  • Search Results

  • sodiumtetraborate20122133043411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium tetraborate

2 Products
Synonym:   Borax, fused
Image of Sodium tetraborate
CAS Number:
Linear Formula:
 Na2B4O7
Molecular Weight:
 201.22
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More