• USA Home
  • Search Results

  • sodiumtetraboratedecahydrate38137130396411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium tetraborate decahydrate

5 Products
Synonym:   Borax decahydrate, Sodium borate decahydrate
Image of Sodium tetraborate decahydrate
CAS Number:
Linear Formula:
 Na2B4O7 · 10H2O
Molecular Weight:
 381.37
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Purity Grade
  Restriction
  pH Value
  Special Grade
Show More