• USA Home
  • Search Results

  • sodiumtetrachloroaurateiiidihydrate397801387402711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium tetrachloroaurate(III) dihydrate

1 Products
Synonym:   Sodium gold chloride
Image of Sodium tetrachloroaurate(III) dihydrate
CAS Number:
Linear Formula:
 NaAuCl4 · 2H2O
Molecular Weight:
 397.80
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More