• USA Home
  • Search Results

  • sodiumtetrakis35bistrifluoromethylphenylborate886207906088111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

2 Products
Synonym:   Benzene, 1,3-bis(trifluoromethyl)-, boron complex, NaBARF, Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron sodium
Image of Sodium tetrakis[3,5-<WBR>bis(trifluoromethyl)<WBR>phenyl]<WBR>borate
CAS Number:
Empirical Formula (Hill Notation):
 C32H12BF24Na
Molecular Weight:
 886.20
 Beilstein/REAXYS Number:
 5474788
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More