• USA Home
  • Search Results

  • sodiumthiophenolate1321693069811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium thiophenolate

1 Products
Synonym:   Benzenethiol sodium salt, Sodium thiophenoxide, Thiophenol sodium salt
Image of Sodium thiophenolate
CAS Number:
Linear Formula:
 C6H5SNa
Molecular Weight:
 132.16
 Beilstein/REAXYS Number:
 3597302
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More