• USA Home
  • Search Results

  • sodiumthiosulfate15811777298711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium thiosulfate

6 Products
Image of Sodium thiosulfate
CAS Number:
Linear Formula:
 Na2S2O3
Molecular Weight:
 158.11
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Special Grade
  pH Value
  Physical Form
Show More