• USA Home
  • Search Results

  • sodiumthiosulfatesolution15811777298711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium thiosulfate solution

2 Products
Image of Sodium thiosulfate solution
CAS Number:
Linear Formula:
 Na2S2O3
Molecular Weight:
 158.11
 MDL number:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Restriction
  Application(s)
  Molecular weight
  Physical Form
Show More