• USA Home
  • Search Results

  • sodiumtrichloroacetate1853765051111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium trichloroacetate

1 Products
Synonym:   NaTCA, TCA sodium salt, Trichloroacetic acid sodium salt
Image of Sodium trichloroacetate
CAS Number:
Linear Formula:
 CCl3CO2Na
Molecular Weight:
 185.37
 Beilstein/REAXYS Number:
 3773249
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More