• USA Home
  • Search Results

  • solventgreen34184912880311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Solvent Green 3

2 Products
Synonym:   D & C Green No. 6
Image of Solvent Green 3
CAS Number:
Empirical Formula (Hill Notation):
 C28H22N2O2
Molecular Weight:
 418.49
 Colour Index Number:
 61565
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More