• USA Home
  • Search Results

  • sophoricoside4323815295411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sophoricoside

2 Products
Synonym:   5′,7′-Dihydroxy-4′-glucosyloxyisoflavone, Genistein-4′-O-glucoside
Image of Sophoricoside
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O10
Molecular Weight:
 432.38
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More