• USA Home
  • Search Results

  • sorbicacid1121311044111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sorbic acid

8 Products
Synonym:   2,4-Hexadienic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H8O2
Molecular Weight:
 112.13
 EC Index Number
 203-768-7
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Mode of Action
  Anal. Standard Form
  Activity Spectrum
  Physical Form
  Assay
Show More