• USA Home
  • Search Results

  • sotalolhydrochloride3088295924011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Sotalol hydrochloride

4 Products
Synonym:   N-[4-[1-Hydroxy-2-(isopropylamino)ethyl]phenyl]methanesulfonamide hydrochloride
Image of (±)-Sotalol hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H20N2O3S · HCl
Molecular Weight:
 308.82
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Application(s)
  Manufacturer designation
  Assay
Show More