• USA Home
  • Search Results

  • spectinomycindihydrochloridepentahydrate495352218932811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Spectinomycin dihydrochloride pentahydrate

4 Products
Image of Spectinomycin dihydrochloride pentahydrate
CAS Number:
Empirical Formula (Hill Notation):
 C14H24N2O7 · 2HCl · 5H2O
Molecular Weight:
 495.35
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Product Line
  Special Grade
  Titertest
  Assay
  Potency
  Color
  Biological Source
  Mode of Action
  Physical Form
  Solubility
Show More