• USA Home
  • Search Results

  • sperminenitricoxidecomplex2623513658713811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Spermine–Nitric oxide complex

1 Products
Synonym:   N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl]-1,3-propanediamine, SPER/NO, Spermine NONOate
Image of Spermine–Nitric oxide complex
CAS Number:
Empirical Formula (Hill Notation):
 C10H26N6O2
Molecular Weight:
 262.35
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More