• USA Home
  • Search Results

  • spironolactone416575201711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Spironolactone

4 Products
Synonym:   4-Pregnen-21-oic acid-17α-ol-3-one-7α-thiol γ-lactone 7-acetate, 7α-(Acetylthio)-17α-hydroxy-3-oxopregn-4-ene-21-carboxylic acid γ-lactone
Image of Spironolactone
CAS Number:
Empirical Formula (Hill Notation):
 C24H32O4S
Molecular Weight:
 416.57
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More