• USA Home
  • Search Results

  • spmefiberassortmentkit51234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

SPME fiber assortment kit 5

2 Products
 NACRES
 NB.21
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Miscellaneous Specifications
  Application(s)
  Size designation
  Material
  Compatibility
  Matrix active group
Show More