• USA Home
  • Search Results

  • squalene4107211102411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Squalene

4 Products
Synonym:   2,6,10,15,19,23-Hexamethyl-2,6,10,14,18,22-tetracosahexaene
Image of Squalene
CAS Number:
Linear Formula:
 [(CH3)2C[=CHCH2CH2C(CH3)]2=CHCH2-]2
Molecular Weight:
 410.72
 Beilstein/REAXYS Number:
 1728919
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More