• USA Home
  • Search Results

  • srnoscapine4134212862111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S,R)-Noscapine

1 Products
Image of (<I>S,R</I>)-Noscapine
CAS Number:
Empirical Formula (Hill Notation):
 C22H23NO7
Molecular Weight:
 413.42
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
363960 (S,R)-Noscapine, 97%,