• USA Home
  • Search Results

  • ssbenzoicacid122126585011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

SS Benzoic Acid

1 Products
Synonym:   Benzoic acid solution
Image of SS Benzoic Acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H6O2
Molecular Weight:
 122.12
 EC Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More