• USA Home
  • Search Results

  • ssdibenzyltrithiocarbonate290472650429011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

S,S-Dibenzyl trithiocarbonate

1 Products
Synonym:   Carbonic acid, trithio-, dibenzyl ester, Carbonotrithioic acid bis(phenylmethyl) ester, DBTTC, Dibenzyl carbonotrithioate, NSC 33081
Image of S,S-Dibenzyl trithiocarbonate
CAS Number:
Empirical Formula (Hill Notation):
 C15H14S3
Molecular Weight:
 290.47
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More