• USA Home
  • Search Results

  • staurosporinefromstreptomycessp466536299674111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Staurosporine from Streptomyces sp.

2 Products
Synonym:   Antibiotic AM-2282, Staurosporine
Image of Staurosporine from <I>Streptomyces</I> sp.
CAS Number:
Empirical Formula (Hill Notation):
 C28H26N4O3
Molecular Weight:
 466.53
 Beilstein/REAXYS Number:
 1060573
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Mode of Action
  Activity Spectrum
  Assay
Show More