• USA Home
  • Search Results

  • stearoylchloride3029211276511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Stearoyl chloride

2 Products
Synonym:   Octadecanoyl chloride
Image of Stearoyl chloride
CAS Number:
Linear Formula:
 CH3(CH2)16COCl
Molecular Weight:
 302.92
 Beilstein/REAXYS Number:
 639784
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 39050509
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More