• USA Home
  • Search Results

  • stearylalcohol2704911292511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Stearyl alcohol

4 Products
Synonym:   Stearyl alcohol
Image of Stearyl alcohol
CAS Number:
Empirical Formula (Hill Notation):
 C18H38O
Molecular Weight:
 270.49
 EC Index Number:
 204-017-6
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
  Anal. Standard Form
  Physical Form
Show More