• USA Home
  • Search Results

  • stearylstearate53696277896311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Stearyl stearate

2 Products
Synonym:   Stearic acid stearyl ester
Image of Stearyl stearate
CAS Number:
Linear Formula:
 CH3(CH2)16COO(CH2)17CH3
Molecular Weight:
 536.96
 Beilstein/REAXYS Number:
 1807499
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Functional group
  Application(s)
Show More