• USA Home
  • Search Results

  • stertbutylthioacetate1322299990611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

S-tert-Butyl thioacetate

1 Products
Image of <I>S</I>-<I>tert</I>-Butyl thioacetate
CAS Number:
Linear Formula:
 CH3COSC(CH3)3
Molecular Weight:
 132.22
 Beilstein/REAXYS Number:
 1740436
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More