• USA Home
  • Search Results

  • stevioside804875781789711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Stevioside

4 Products
Synonym:   (4α)-13-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid β-D-glucopyranosyl ester, Steviosin
Image of Stevioside
CAS Number:
Empirical Formula (Hill Notation):
 C38H60O18
Molecular Weight:
 804.87
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Shelf Life
  Application(s)
  Assay
  Special Grade
Show More