• USA Home
  • Search Results

  • streptomycinsulfate72869381074011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Streptomycin sulfate

3 Products
Synonym:   Streptomycin sulfate salt
Image of Streptomycin sulfate
CAS Number:
Linear Formula:
 C21H39N7O12 · 1.5 H2SO4
Molecular Weight:
 728.69
 Beilstein/REAXYS Number:
 3894995
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Manufacturer designation
Show More