• USA Home
  • Search Results

  • streptonigrinfromstreptomycesflocculus50646393019611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Streptonigrin from Streptomyces flocculus

1 Products
Synonym:   Bruneomycin, Nigrin
Image of Streptonigrin from <I>Streptomyces flocculus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C25H22N4O8
Molecular Weight:
 506.46
 Beilstein/REAXYS Number:
 599390
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More