• USA Home
  • Search Results

  • strontiumnitrate211631004276911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Strontium nitrate

4 Products
Image of Strontium nitrate
CAS Number:
Linear Formula:
 Sr(NO3)2
Molecular Weight:
 211.63
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Purity Grade
  Potency
  Melting Point
  Density
  Bulk Density
  Loss
  Physical Form
  Special Grade
  pH Value
Show More