• USA Home
  • Search Results

  • strontiumtitanate183491206059211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Strontium titanate

3 Products
Synonym:   Strontium titanium trioxide
Image of Strontium titanate
CAS Number:
Linear Formula:
 SrTiO3
Molecular Weight:
 183.49
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Semiconductor properties
  Hardness
  Composite Dimension
  Physical Form
  Dielectric constant
  Assay
Show More