• USA Home
  • Search Results

  • strychninehydrochloride37087142186911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Strychnine hydrochloride

1 Products
Image of Strychnine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H22N2O2 · HCl
Molecular Weight:
 370.87
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
S8753 Strychnine hydrochloride,