• USA Home
  • Search Results

  • styrene1041510042511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Styrene

4 Products
Synonym:   Phenylethylene, Vinylbenzene
Image of Styrene
CAS Number:
Linear Formula:
 C6H5CH=CH2
Molecular Weight:
 104.15
 Beilstein/REAXYS Number:
 1071236
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Autoignition temp
  Additive
  Physical Form
  Application(s)
  Assay
Show More