• USA Home
  • Search Results

  • styrened8112201936162711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Styrene-d8

1 Products
Synonym:   Deuterated styrene, Octadeuterostyrene, Phenylethylene-D8, Vinylbenzene-D8
Image of Styrene-d<SUB>8</SUB>
CAS Number:
Linear Formula:
 C6D5CD=CD2
Molecular Weight:
 112.20
 Beilstein/REAXYS Number:
 2327045
 EC Number:
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More