• USA Home
  • Search Results

  • styreneoxide120159609311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Styrene oxide

1 Products
Synonym:   1,2-Epoxyethylbenzene, Phenylethylene oxide, Phenyloxirane
Image of Styrene oxide
CAS Number:
Empirical Formula (Hill Notation):
 C8H8O
Molecular Weight:
 120.15
 Beilstein/REAXYS Number:
 108582
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
S5006 Styrene oxide, 97%,