• USA Home
  • Search Results

  • styrenesolution1041510042511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Styrene solution

1 Products
Synonym:   Phenylethylene
Image of Styrene solution
CAS Number:
Empirical Formula (Hill Notation):
 C8H8
Molecular Weight:
 104.15
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More