• USA Home
  • Search Results

  • succinicacid1180911015611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Succinic acid

18 Products
Synonym:   Butanedioic acid, Succinic acid
Image of Succinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C4H6O4
Molecular Weight:
 118.09
 EC Index Number
 203-740-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  Anal. Standard Form
  Product Line
  Biological Source
  Greener Alternative Principles
  Analyte Suitability
  Physical Form
  pH Value
  Transition temperature
  Potency
  Density
  Bulk Density
  Autoignition temp
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Purity Grade
  Melting Point
  Special Grade
  Solubility
  Assay
Show More

Product #
Description
S3674 Succinic acid, BioXtra, ≥99.0%, Green Alternative
398055 Succinic acid, ACS reagent, ≥99.0%,
S7501 Succinic acid, ReagentPlus®, ≥99.0%, Green Alternative
S9512 Succinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, Green Alternative
14079 Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T),
14080 Succinic acid, purum p.a., ≥99.0% (T),
W502715 Succinic acid, natural,
69612 Succinic acid, matrix substance for MALDI-MS, ≥99.5% (T), Ultra pure,
1.00681 Succinic acid, cryst., EMPROVE® ESSENTIAL NF,JPE,ACS,
1.00682 Succinic acid, for analysis EMSURE® ACS,
W471920 Succinic acid, ≥99%, FCC, FG,
1623411 Succinic acid, United States Pharmacopeia (USP) Reference Standard,
797987 Succinic acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%,
8.22260 Succinic acid, for synthesis,
49893 Succinic acid, certified reference material, TraceCERT®,
W471912 Succinic acid, natural, ≥99%, FG,
Y0001988 Succinic acid, European Pharmacopoeia (EP) Reference Standard,
PHR1418 Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material,