• USA Home
  • Search Results

  • succinicaciddisodiumsalt1620515090311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Succinic acid disodium salt

1 Products
Synonym:   Sodium succinate dibasic
Image of Succinic acid disodium salt
CAS Number:
Linear Formula:
 NaOOCCH2CH2COONa
Molecular Weight:
 162.05
 Beilstein/REAXYS Number:
 3917246
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More