• USA Home
  • Search Results

  • sucralose397635603813211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sucralose

5 Products
Synonym:   1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose
Image of Sucralose
CAS Number:
Empirical Formula (Hill Notation):
 C12H19Cl3O8
Molecular Weight:
 397.63
 Beilstein/REAXYS Number:
 3654410
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Titertest
  Assay
  Special Grade
  Anal. Standard Form
Show More