• USA Home
  • Search Results

  • sucrosesaccharose342305750111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sucrose (saccharose)

2 Products
Synonym:   α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Saccharose, Sucrose, Sugar
Image of Sucrose (saccharose)
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11
Molecular Weight:
 342.30
 Beilstein/REAXYS Number:
 90825
 EC Index Number:
 200-334-9
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  pH Value
  Potency
  Bulk Density
Show More