• USA Home
  • Search Results

  • sudanredg27831122955611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sudan red G

2 Products
Synonym:   1-(2-Methoxyphenylazo)-2-naphthol, Oil Red 113
Image of Sudan red G
CAS Number:
Empirical Formula (Hill Notation):
 C17H14N2O2
Molecular Weight:
 278.31
 Beilstein/REAXYS Number:
 1843558
 Colour Index Number:
 12150
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More