• USA Home
  • Search Results

  • sulfadiazine250286835911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfadiazine

8 Products
Synonym:   4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, N1-(Pyrimidin-2-yl)sulfanilamide
Image of Sulfadiazine
CAS Number:
Empirical Formula (Hill Notation):
 C10H10N4O2S
Molecular Weight:
 250.28
 Beilstein/REAXYS Number:
 6733588
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Shelf Life
  Titertest
  Mode of Action
  Anal. Standard Form
  Physical Form
  Special Grade
Show More