• USA Home
  • Search Results

  • sulfadiazinesodiumsalt2722654732011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfadiazine sodium salt

1 Products
Synonym:   4-Amino-N-(2-pyrimidinyl)benzenesulfonamide sodium salt, SPS Agar Supplement
Image of Sulfadiazine sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C10H9N4NaO2S
Molecular Weight:
 272.26
 Beilstein/REAXYS Number:
 4068213
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More