• USA Home
  • Search Results

  • sulfadimidineforpeakidentification278335768111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfadimidine for peak identification

1 Products
Synonym:   4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine, Sulfamethazine
Image of Sulfadimidine for peak identification
CAS Number:
Empirical Formula (Hill Notation):
 C12H14N4O2S
Molecular Weight:
 278.33
 Beilstein/REAXYS Number:
 261304
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More