• USA Home
  • Search Results

  • sulfadoxine31033244757611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfadoxine

2 Products
Synonym:   4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide, Sulfadoxin, Sulfadoxine
Image of Sulfadoxine
CAS Number:
Empirical Formula (Hill Notation):
 C12H14N4O4S
Molecular Weight:
 310.33
 Beilstein/REAXYS Number:
 625453
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More