• USA Home
  • Search Results

  • sulfameter2803065106911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfameter

2 Products
Synonym:   5-Methoxysulfadiazine, N1-(5-Methoxypyrimidin-2-yl)sulfanilamide, Sulfamethoxydiazine
Image of Sulfameter
CAS Number:
Empirical Formula (Hill Notation):
 C11H12N4O3S
Molecular Weight:
 280.30
 Beilstein/REAXYS Number:
 621130
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Application(s)
Show More