• USA Home
  • Search Results

  • sulfamethazine278335768111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfamethazine

3 Products
Synonym:   4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine
Image of Sulfamethazine
CAS Number:
Empirical Formula (Hill Notation):
 C12H14N4O2S
Molecular Weight:
 278.33
 Beilstein/REAXYS Number:
 261304
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Special Grade
  Titertest
  Assay
  Mode of Action
Show More