• USA Home
  • Search Results

  • sulfamicacid9709532914611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfamic acid

9 Products
Synonym:   Amidosulfonic acid
Image of Sulfamic acid
CAS Number:
Linear Formula:
 NH2SO3H
Molecular Weight:
 97.09
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  pH Value
  Application(s)
  Melting Point
  Ice Shipping
  Solubility
  Special Grade
Show More