• USA Home
  • Search Results

  • sulfanilicacid1731912157311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfanilic acid

8 Products
Synonym:   4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid
Image of Sulfanilic acid
CAS Number:
Linear Formula:
 4-(H2N)C6H4SO3H
Molecular Weight:
 173.19
 Beilstein/REAXYS Number:
 908765
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Anal. Standard Form
  pH Value
  Application(s)
  Potency
  Melting Point
  Bulk Density
  Autoignition temp
  Special Grade
  Solubility
  Density
  Physical Form
Show More