• USA Home
  • Search Results

  • sulfanilicacidazochromotrop570412364714511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfanilic acid azochromotrop

1 Products
Synonym:   1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6-disulfonic acid trisodium salt, 2-(4-Sulfophenylazo)-1,8-dihydroxy-3,6-naphthalene disulfonic acid trisodium salt, 2-(4-Sulfophenylazo)chromotropic acid trisodium salt, SPADNS
Image of Sulfanilic acid azochromotrop
CAS Number:
Empirical Formula (Hill Notation):
 C16H9N2Na3O11S3
Molecular Weight:
 570.41
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More